Learn More
Memmert™ Ethernet Connection Cable
Product Code. 17140185 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
ICO105
ICO150
ICO240
ICO50
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17140185
Brand: Memmert™ IPP260ecoE06189
Only null left
Add to basket
Add to basket
5 m ethernet connection cable for computer interface
  • For computer interface
  • Accessory
  • Notification by SMS in case of any error or alarm of the device
Specifications
For Use With (Equipment) IPP260eco
Length (Metric) 5 m