Learn More
Tocris Bioscience™ ET 18-OCH3
Selective PI-PLC inhibitor, also PAF receptor agonist
Brand: Tocris Bioscience™ 7462/10
This item is not returnable.
View return policy
Description
ET 18-OCH3 is a synthetic lysophospholipid analog that selectively inhibits phosphatidylinositol phospholipase C (IC50 = 9.6 μM in fibroblasts and adenocarcinoma cells). ET 18-OCH3 acts as an agonist at platelet-activating factor (PAF) receptors. ET 18-OCH3 is an antitumor lipid that selectively induces apoptosis in tumor cells, sparing normal cells, and activates the intracellular Fas/CD95 death receptor. ETH 18-OCH3 also triggers eryptosis and interacts with mitochondrial membranes to increase mitochondrial membrane permeability, leading to dysfunction and apoptosis. Racemic mixture.Specifications
4-Hydroxy-7-methoxy-N,N,N-trimethyl-3,5,9-trioxa-4-phosphaheptacosan-1-aminium 4-oxide | |
10 mg | |
C27H58NO6P |
70641-51-9 | |
Phospholipase C Inhibitors |