missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Escitalopram oxalate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Escitalopram oxalate is a selective 5-HT reuptake inhibitor (Ki values are 0.89, 8150, and 10500 nM for serotonin, noradrenalin, and dopamine transporters respectively). Escitalopram oxalate is an S-enantiomer and eutomer of citalopram (Cat. No. 1427).
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 414.43 |
Percent Purity | 98% |
Chemical Name or Material | Escitalopram oxalate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction