missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ES Type 9 Tag Out
Brand: Brady™ 284428
This item is not returnable.
View return policy
Specifications
ES Type 9 Tag Out | |
Polyester | |
5.71 in. | |
10 Pk. | |
-40°C to +100°C | |
3.35 in. |
Black/Yellow on White | |
Safety Identification | |
145 mm | |
-40°F to +212°F | |
0.249 mm | |
85 mm |