Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks With Conical Ground Joint
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03022949
Product Code. 10230931
  • 18.00€ / Each
Edge
Explore more special offers
Description
Description
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
DIN EN ISO 4797
Borosilicate Glass, Duran Glass
NS29/32
250 mL
With scale (approx. volume/mL), with inscription label