missing translation for 'onlineSavingsMsg'
Learn More

ER 27319 maleate, Tocris Bioscience™

Product Code. 18045562
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective Syk kinase inhibitor

Selective inhibitor of Syk kinase. Inhibits tyrosine phosphorylation of Syk initiated by the engagement of FcεRI in rat and human mast cells. Results in the abrogation of degranulation, TNF-α production (IC50 = 10 μM) and other related signaling events.

Chemical Identifiers

CAS 1204480-26-1
Molecular Formula C22H24N2O5
Molecular Weight (g/mol) 396.443
InChI Key WVUQPGFRFBVJKH-BTJKTKAUSA-N
Synonym er 27319 maleate, 10-3-aminopropyl-3,4-dimethyl-9 10h-acridinone maleate
PubChem CID 56972172
IUPAC Name 10-(3-aminopropyl)-3,4-dimethylacridin-9-one;(Z)-but-2-enedioic acid
SMILES CC1=C(C2=C(C=C1)C(=O)C3=CC=CC=C3N2CCCN)C.C(=CC(=O)O)C(=O)O

Specifications

Quantity 50 mg
Formula Weight 396.17
Percent Purity >99%
Chemical Name or Material ER 27319 maleate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.