Learn More
Eppendorf™ epT.I.P.S.™ Sets - Pipette Tips
GREENER_CHOICE
Eppendorf epT.I.P.S.™ Sets, in 'Eppendorf Quality' grade, combine the convenience of a reusable Box 2.0 with high-quality reloadable pipette tips, offering a sustainable, efficient solution for precise and organized pipetting in laboratories.
Brand:  Eppendorf™ 0030076338
59.85 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 17392900
  • 62.20€ / Pack of 480
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • The Reloads and Boxes can be autoclaved at 121 °C for subsequent use
  • epT.I.P.S. Box 2.0 and epT.I.P.S. Set available in Eppendorf Quality
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 to 300 μL
Non-sterile
Autoclavable
Eppendorf Quality (Non-sterile)
Box
480 Pk.
Orange
Air-cushion System
epT.I.P.S.
Non-filtered
Colorless
epTIPS
Eppendorf Pipettes, Non-Eppendorf Pipettes
480 Tips (5 Trays x 96 Tips), 1 Reusable Box
Pipette Tips
Single-use
Yes
Polypropylene