missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ PCR Plates 384-well, 45 μL - PCR Plates
GREENER_CHOICE

Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.

Brand:  Eppendorf™ 0030129350

131.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10267467

Quantity Price
1 138.00€ / Pack of 10
5 127.17€ / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket

This item is not returnable. View return policy

Description

Description

  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean).
  • Safe Code option provides 3-level coding for safe sample and vessel identification.
  • ECC 200 error correction function enables safe code reading with up to 30% damaged codes.

Certifications

Certified free of human DNA, DNase, RNase, and PCR inhibitors

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Barcode
Standard
Autoclavable
Blue
Automated Systems
Polypropylene, Polycarbonate
384
Round Bottom (U-well)
PCR Clean (Non-sterile)
Colorless
Free of Human DNA, DNase, RNase and PCR Inhibitors
16 x 24
10 x Individually Blister Wrapped
PCR Plate
skirted
45 μL
Non-sterile
2250 x g
10 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions