missing translation for 'onlineSavingsMsg'
Learn More

Epcam / CD326 Mouse, Clone: EGP40/1373, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 4072-MSM13-P1

 View more versions of this product

Product Code. 15920418

  • 510.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Epcam / CD326
Cell surface
EGP40/1373
Unconjugated
NP_002345
Mouse
40-43kDa
Protein A/G Purified
RUO
4072
IgG2b
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
EPCAM
Recombinant human EpCAM protein fragment from the extracellular domain (aa77-202)
HT29 cells or Ovarian carcinoma
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions