Learn More
Enzymaster Enzyme Screening Kit
The Enzyme Screening Kit contains a wide range of wild type and engineered enzymes with a broad substrate scope for your screening!
Brand: Enzymaster 2004-00
This item is not returnable.
View return policy
Description
- Ketoreductase Kit
- Enzyme kit containing 20 ketoreductases (E.C. 1.1.1.) for a quick screening
- Applicable for ketone reduction and alcohol oxidation
- Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
- 0.5 g per enzyme
- Formulated as lyophilized cell-free extract (CFE)
- IRED Kit
- Enzyme kit containing 16 iminereductases (E.C. 1.5.1.) for a quick screening
- Applicable for imine reduction and reductive amination
- Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
- 0.5 g per enzyme
- Formulated as lyophilized cell-free extract (CFE)
- Alolase Kit
- Enzyme kit containing 20 aldolases (E.C. 4.1.2.) for a quick screening
- Contains threonine, phenylserine and serine aldolases
- Applicable for reversible aldol reaction of glycine and an aldehyde to β-hydroxy-α-l-amino acid
- Diverse range of substrates possible, high selectivity
- 0.5 g per enzyme
- Formulated as lyophilized cell-free extract (CFE)
- Transaminase Kit
- Enzyme kit containing 21 transaminases (E.C. 2.6.1.) for a quick screening
- Applicable for reversible transamination of an amino group from an amino donor to a keto amino acceptor
- Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
- 0.5 g enzyme powder per vial
- Formulated as lyophilized cell-free extract (CFE)
Specifications
Enzyme Screening Kit | |
21 vials with 0.5 g enzyme each as lyophilized cell-free extract (CFE) | |
Enzyme screening of transaminases |