missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enzymaster Enzyme Screening Kit
Click to view available options
For Use With (Application):
Enzyme screening of aldolases
Enzyme screening of iminereductases
Enzyme screening of ketoreductases
Enzyme screening of transaminases
Kit Contents:
16 vials with 0.5 g enzyme (CFE)
20 vials with 0.5 g enzyme (CFE)
21 vials with 0.5 g enzyme (CFE)
Unit Size:
Each
Description
- Ketoreductase Kit
- Enzyme kit containing 20 ketoreductases (E.C. 1.1.1.) for a quick screening
- Applicable for ketone reduction and alcohol oxidation
- Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
- 0.5 g per enzyme
- Formulated as lyophilized cell-free extract (CFE)
- IRED Kit
- Enzyme kit containing 16 iminereductases (E.C. 1.5.1.) for a quick screening
- Applicable for imine reduction and reductive amination
- Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
- 0.5 g per enzyme
- Formulated as lyophilized cell-free extract (CFE)
- Alolase Kit
- Enzyme kit containing 20 aldolases (E.C. 4.1.2.) for a quick screening
- Contains threonine, phenylserine and serine aldolases
- Applicable for reversible aldol reaction of glycine and an aldehyde to β-hydroxy-α-l-amino acid
- Diverse range of substrates possible, high selectivity
- 0.5 g per enzyme
- Formulated as lyophilized cell-free extract (CFE)
- Transaminase Kit
- Enzyme kit containing 21 transaminases (E.C. 2.6.1.) for a quick screening
- Applicable for reversible transamination of an amino group from an amino donor to a keto amino acceptor
- Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
- 0.5 g enzyme powder per vial
- Formulated as lyophilized cell-free extract (CFE)
Specifications
Specifications
| Product Type | Enzyme Screening Kit |
| Kit Contents | 20 vials with 0.5 g enzyme (CFE) |
| For Use With (Application) | Enzyme screening of aldolases |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction