Learn More
Enzymaster Enzyme Screening Kit
The Enzyme Screening Kit contains a wide range of wild type and engineered enzymes with a broad substrate scope for your screening!
Brand:  Enzymaster 2002-00
 View more versions of this product
Product Code. 17874621
  • 8235.00€ / Each
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Description
  • Ketoreductase Kit
    • Enzyme kit containing 20 ketoreductases (E.C. 1.1.1.) for a quick screening
    • Applicable for ketone reduction and alcohol oxidation
    • Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
    • 0.5 g per enzyme
    • Formulated as lyophilized cell-free extract (CFE)
  • IRED Kit
    • Enzyme kit containing 16 iminereductases (E.C. 1.5.1.) for a quick screening
    • Applicable for imine reduction and reductive amination
    • Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
    • 0.5 g per enzyme
    • Formulated as lyophilized cell-free extract (CFE)
  • Alolase Kit
    • Enzyme kit containing 20 aldolases (E.C. 4.1.2.) for a quick screening
    • Contains threonine, phenylserine and serine aldolases
    • Applicable for reversible aldol reaction of glycine and an aldehyde to β-hydroxy-α-l-amino acid
    • Diverse range of substrates possible, high selectivity
    • 0.5 g per enzyme
    • Formulated as lyophilized cell-free extract (CFE)
  • Transaminase Kit
    • Enzyme kit containing 21 transaminases (E.C. 2.6.1.) for a quick screening
    • Applicable for reversible transamination of an amino group from an amino donor to a keto amino acceptor
    • Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
    • 0.5 g enzyme powder per vial
    • Formulated as lyophilized cell-free extract (CFE)
Specifications
Specifications
Enzyme Screening Kit
16 vials with 0.5 g enzyme each as lyophilized cell-free extract (CFE)
Enzyme screening of iminereductases