Learn More
Cole-Parmer™ Environmental Notebook
Brand:  Cole-Parmer™ 06570-00
32.59 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Product Code. 15199923
  • 41.45€ / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Specifications
4.75 x 7.5 in.
156
Environmental Notebook
1