Learn More
Memmert™ Entry Port 23 mm for UF55, Standard Position Right Side Center/Center
Optional entry port for Memmert Ovens.
Brand: Memmert™ UF55F2
106.86 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
- Entry port with 23 mm diameter
- Flap and stopper closure
- Standard position right side center/center
Specifications
UF55 Oven | |
Entry Port | |
23 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Promotions
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm for UF55, Standard Position Right Side Center/Center > For Use With: UF55