Learn More
Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for the iCAP™ Series ICP-OES Systems
Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance plasma torch for iCAP™ Series ICP-OES systems.
392.00€
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12794868
|
Thermo Scientific™
842312051841 |
EMT Duo Torch for iCAP 7000 Series ICP-OES systems |
392.00€
Each |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for Thermo Scientific™ iCAP™ Series ICP-OES systems. The torch clips in and can be removed in just seconds.
Enhanced performance through advanced accessoriesWe offer dedicated torch designs for radial and axial plasma views, including:
- EMT Radial Torch for radial-only systems
- EMT Duo Torch for radial and axial (duo) view systems