Learn More
Corning™ Endothelial Cell Growth Supplement (ECGS)
GREENER_CHOICE
Product Code. p-90818 Shop All Corning Products
Click to view available options
Quantity:
15 mg
100 mg
Unit Size:
100mg
15mg
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 11523610
Brand: Corning™ 356006
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Ideal for endothelial cell culture
The Corning portfolio of cytokines, growth factors, and media supplementation offers a wide array of proteins that result in proliferation or differentiation of the affected cells.
  • Vialed, highly purified growth factors with high biological activity
  • Broad range of highly purified natural and recombinant lymphokines
  • Enriched culture supplements to be reconstituted/added in the medium of your choice
  • Propagate your cells under more defined serum-reduced or serum-free conditions
Corning offers a broad range of other media growth supplements. Some products are specific to cell type, such as SMC4 (for iPSCs) and T-Cell culture supplement (for T-cells), while other products promote proliferation in a wide variety of cell types. Reducing serum use is made easy with products such as ITS and Corning Nu-SerumTM.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2° to 8°C
Description ECGS is a broadly used supplement to culture a variety of cells, particularly endothelial cells. ECGS contains various growth factors (e.g., acidic FGF or ECGF-a)
Concentration 50 to 150 µg/ mL depending on cell type. May require supplementation of 1 to 100 µg/mL heparin.
Product Type Growth Factor
Formulation Lyophilized from NaCl-containing buffer with streptomycin sulfate; reconstitute in serum-free medium
Quantity 100 mg
Source Bovine brain
Tested For Tested for ability to promote proliferation of Fetal Bovine Heart Endothelial Cells (FBHEC) in medium containing 10% newborn calf serum. Filtered (0.2 µm membrane) and tested and found negative for bacteria, fungi and mycoplasma.
compliance-icons compliance-icons
Product Identifier
  • 100MG Endothelial Cell Growth Supplement (ECGS)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Streptomycin sulfate