Learn More
Merck Millicell™ EZ Slides
Shop All Merck Products
Click to view available options
Culture Area:
0.7 cm2/well
1.7 cm2/well
Volume (Metric) Working:
0.2 to 0.7 mL/Well
0.5 to 1.7 mL/Well
Wells:
4
8
Unit Size:
Pack of 16
Pack of 96
Description
- Polypropylene wells are resistant to most fixatives—cells may be fixed and stained with wells still attached
- Breakable tabs for easy well removal—no tools required to remove wells from slide
- No adhesive used to attach wells to slide
- No leaky wells
- Raised corner ridges on lid facilitate stacking of multiple units in the incubator
- Noncytotoxic and nonpyrogenic
- Sterile
- Slide holder (available separately) protects slide from damage
Specifikationer
Specifikationer
| Type | EZ Slide |
| Wells | 8 |
| Color | Clear |
| Well Material | Polypropylene |
| Culture Area | 0.7 cm2/well |
| Lid Material | Polystyrene |
| Material | Glass |
| Volume (Metric) Working | 0.2 to 0.7 mL/Well |
| For Use With (Application) | Simplify your cell analysis by using the Millicell EZ SLIDE glass slide to culture, fix, stain and view your sample all in one device |
| Sterility | Sterile |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering