missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Elinogrel
P2Y12 antagonist
213.00€ - 850.00€
Specifications
Purity | 97% |
---|---|
Content And Storage | -20°C |
Product Type | Elinogrel |
Molecular Weight (g/mol) | 523.95 |
Inhibitors | Elinogrel |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18770769
|
Tocris Bioscience™
5316/50 |
50 mg |
850.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18766248
|
Tocris Bioscience™
5316/10 |
10 mg |
213.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P2Y12 antagonist; exhibits antiplatelet activity.Specifications
97% | |
Elinogrel | |
Elinogrel |
-20°C | |
523.95 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title