missing translation for 'onlineSavingsMsg'
Learn More

Multi 3 ions, 20mg/L of each Component, For Ion Chromatography, 0.1% NaOH matrix, Fisher Chemical™

Product Code. 12952235 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 12952235

Brand: Fisher Chemical A 102 100

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Chemical Name or Material Multi 3 ions
Packaging Bottle
Analyte or Component Names Bromates (BrO3-), Chlorates (ClO3-), Iodates (IO3)
Physical Form Liquid
Concentration 20 mg/L of each Component
Type Multi-component Standard Solution
Element Bromates, Chlorates, Iodates
Solvent or Matrix 0.1% NaOH
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.