missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Electronic Decapper
1055.00€
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12838595
|
Macherey-Nagel™
735611 |
For 11mm Aluminum Crimp Caps |
1055.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12848595
|
Macherey-Nagel™
735620 |
For 20mm Aluminum Crimp Caps |
1055.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Electronic Decapper | |
1 Each |