Learn More
Fisherbrand™ Electronic Crimping Tools, Handheld, Battery
Product Code. p-7113953 Shop All Fisherbrand Products
Click to view available options
Description:
11mm Electronic Crimper
Unit Size:
Each
Product Code. 15582300
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Reduces hand strain compared to manual crimper operation.
  • Electronic Crimper
  • 110 - 240V
  • Electronic
  • Steel/plastic
  • Crimper
  • Adjustable
  • For Aluminum/Steel caps
TRUSTED_SUSTAINABILITY
Specifications
Description 11mm Electronic Crimper
Diameter (Metric) Cap 11 mm
Material Plastic, Steel