Learn More
WTW™ Electrolyte for WTW™ SenTix™ ORP Electrode
Brand:  WTW™ 109735
Product Code. 11702155
  • 47.15€ / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
118°C
Clear
Combined SenTix™ ORP Electrode with Ag-Electrode
Electrolyte
-14°C
7 (at 25°C)
250 mL
2 mol/L KNO3, 0.001 mol/L KCl
Electrolyte
2mol/L KNO3 0.001mol/L KCl, bottle with 250mL
Bottle
Yes