missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EED 226

Potent and selective EED inhibitor

181.00€ - 694.00€

Specifications

Chemical Name or Material N-(2-Furanylmethyl)-8-[4-(methylsulfonyl)phenyl]-1,2,4-triazolo[4,3-c]pyrimidin-5-amine
CAS 2083627-02-3
Target EED
Molecular Formula C17H15N5O3S
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18727786
View Documents
Tocris Bioscience™
7762/10
10 mg
181.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
18719266
View Documents Promotion Details
Tocris Bioscience™
7762/50
50 mg
694.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EED 226 is a potent (IC50 = 23.4 nM) and selective EED inhibitor. It inhibits PRC2 activity by binding to the H3K27me3 pocket of EED. The compound shows selectivity for PRC2 complex over protein methyltransferases, kinases, and other protein classes. In vitro, EED 226 regulates histone H3K27 methylation and PRC2 target gene expression. In vivo, it induces tumor regression in a mouse xenograft model. EED 226 is orally bioavailable.
Specifications

Specifications

N-(2-Furanylmethyl)-8-[4-(methylsulfonyl)phenyl]-1,2,4-triazolo[4,3-c]pyrimidin-5-amine
2083627-02-3
EED
C17H15N5O3S
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.

SDS
Documents

Documents

Product Certifications