missing translation for 'onlineSavingsMsg'
Learn More

EDTA Disodium Salt Solution, 0.1M, Chem-Lab

Product Code. 10214142
Click to view available options
Quantity:
500 mL
1 L
5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in electrophoresis and molecular biology applications. EDTA Disodium Salt Solution, 0.1M, Chem-Lab is a chelator of divalent cations for use in TH titrations. Solution contains 37.224g EDTA diNa · 2H2O per liter of water (0.01 M). Sol A = 10°FH/mL or 5.6°DH/mL.

EDTA Disodium Salt Dihydrate is a chelator of divalent cations. It inhibits enzymes, such as metalloproteases, that require divalent cations for activity. It is also used in many enzyme buffers and as an enzyme inactivator at high concentrations.

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point 0°C
Color Colorless
Density 1.02g/cm3
pH ±9
Boiling Point 100°C
Odor Odorless
Quantity 1 L
Solubility Information Soluble
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Formula Weight 372.24g/mol
Concentration or Composition (by Analyte or Components) 0.1mol/L (± 0.0004, 20°C)
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.