missing translation for 'onlineSavingsMsg'
Learn More

Ansell EDGE™ 48-126 Polyurethane Coated Seamless Knit Industrial Work Gloves

Product Code. 16875230
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Industrial work gloves feature a seamless knit liner and elasticized knit wrist, with a polyurethane coating for abrasion resistance.

  • An inexpensive, light duty glove with quality features
  • Seamless structure ensures comfort and encourages safety
  • 15-gauge polyester offers flexibility and dexterity
  • High tactility in dry or slightly oily environments
  • Dark color hides dirt for an extended useful life
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Polyurethane
Color Black
Hand Specific
Material Polyester
Product Type Seamless Knit Gloves
Size 11
For Use With (Application) Automotive Assembly, Oil/Gas, and Machinery/Equipment Operation
Glove Cuff Style Knit
Series 48-126
Yarn Gauge 15
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'