Learn More
BTX™ ECM™ 2001 Safety Stand for Flatpack
ECM™ 2001 Safety Stand for Flatpack for use with ECM™ 2001.
Brand:  BTX™ 470208
Product Code. 15467380
  • 2835.00€ / Each
Explore more special offers
Description
Description
  • Cuvette module plugs directly into generator
  • Holds one cuvette
  • Safety swing shield secures cuvette in holder
  • 4 mm Banana Cables, 4 mm F/F Adapters
Specifications
Specifications
Electroporation
BTX Electroporation
Legacy ECM 2001 Safety Stand for Flatpack Chamber
Non-sterile
Digital User
Legacy ECM™ 2001
Electroporation accessory
Safety Stand for Flatpack Only
Banana Cables
AC Sine Wave, DC Square Wave