Learn More
BD Vacutainer Eclipse™ Signal™ Blood Collection Needles
Combination of proven, robust safety technology with improved in-vein confirmation.
Brand: BD Vacutainer 368838
This item is not returnable.
View return policy
Description
- Combination of proven, robust safety technology with the additional benefit of improved in-vein confirmation (speed and visibility of flash) resulting in ease of use and confidence during venous blood collection.
- Minimizes the risk of needle stick injuries during blood collection, thereby increasing both healthcare worker and patient safety.
- Fastest flow among BD straight needles.
Specifications
Black | |
22 G | |
25 mm |