Learn More
Terumo™ Luer-Lock Tip Three-Part Syringe
Terumo™ Luer-Lock Tip Three-Part Syringes are easy to use with their smooth plunger action and have clear barrels to allow quick recognition of air bubbles for more accurate dosing. With easy to read markings, you can be confident you are delivering the intended dose every time.
Brand:  Terumo™ 1SS30LE1
 View more versions of this product
Product Code. 11743323
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
Use these syringes for the aspiration of fluids or sampling blood or other body fluids. Terumo™ Luer-Lock Three-Part Syringes feature a unique double beveled design for greater patient comfort.
  • Silicone coated barrel for a smooth plunger action
  • Clear barrel to allow quick recognition of air bubbles
  • Easy to read markings
  • Latex free
  • Sterile and packed in individually wrapped tamper peel packs
  • With concentric tips
  • Available in a wide range of sizes
Specifications
Specifications
Autoclavable
Polypropylene
For the injection of fluids immediately after filling into the human body, for aspiration of fluids or for the sampling of blood or other body fluids
Luer-Lock Tip Three-Part Syringe, Concentric
50 Pk.
Bevel
Yes
Class IIa, sterile product following MDD 93/42/EEC, CE 0197
Screen printed graduation
Yes
Single-use
30 mL
Luer Lock
Sterile