missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ EC Science Silicon Septa

Designed for long lifetime and low injection port adhesion.

63.35€ - 146.00€

Specifications

Type EC
View More Specs

Products 5
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
15258623
View Documents
Trajan Scientific and Medical™
041912
11 mm This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15218623
View Documents
Trajan Scientific and Medical™
041901
9.5 mm
63.35€
Pack of 25
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
15228623
View Documents
Trajan Scientific and Medical™
041904
11.5 mm
64.60€
Pack of 25
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
15238623
View Documents
Trajan Scientific and Medical™
041905
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
15248623
View Documents
Trajan Scientific and Medical™
041906
12.5 mm
64.60€
Pack of 24
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
 
Description

Description

  • Combines long injection lifetime, low bleed and low injection port adhesion
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications

Specifications

EC
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.