missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Agarose Gels Starter Pak, 1.2% (EU only)

Product Code. 10740584
Click to view available options
Quantity:
Each
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10740584

Brand: Invitrogen™ G600001EU

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

E-Gel™ Agarose Gels Starter Pak, 1.2% (EU only)

E-Gel™ Single Comb Gels are supplied in a 12-well mini-gel format, with a choice of either SYBR Safe™, or ethidium bromide DNA stain. Each gel contains a single row of 12 sample wells. E-Gels are available in two pack sizes-a Starter Pak, and 18-gel multi-paks.
TRUSTED_SUSTAINABILITY

Specifications

Description E-Gel™ Agarose Gels Starter Pak, 1.2% Gel Percentage, EU Plug Type, Single Comb Well Format, 12 Wells, Ethidium Bromide In-Gel Dye, For Use With Nucleic Acid Gel Electrophoresis and Blotting
Concentration 1.2%
Format Single Comb (Well)
Product Type Agarose Gels Starter Pak
For Use With (Application) Nucleic acid gel electrophoresis and blotting
Kit Contents 6 E-Gels, the E-Gel™ PowerBase™ v.4, and a manual. 18-Paks contain 18 gels and a manual
Quantity Each
Storage Requirements Room temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.