Learn More
IBA Lifesciences pASG-IBA E. coli Expression Vector with No Secretion
GREENER_CHOICE
Product Code. p-7139098 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pASG-IBA103
pASG-IBA105
pASG-IBA123
pASG-IBA143
pASG-IBA145
pASG-IBA162
pASG-IBA164
pASG-IBA167
pASG-IBA168
pASG-IBA23
pASG-IBA3
pASG-IBA43
pASG-IBA45
pASG-IBA5
pASG-IBA62
pASG-IBA64
pASG-IBAwt1
Unit Size:
5µg
Product Code. 15603758
Brand: IBA Lifesciences 54164001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Regulate the expression of proteins with a vector that carries the inducible tetracycline promoter/operator. IBA Lifesciences GmbH E. coli Expression Vector with No Secretion allows for the expression of proteins in E.coli.
  • The pASG-IBA vectors are designed for expression of target proteins without affinity tag in E. coli.
  • The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, and the inducible tetracycline promoter/operator for the regulated expression of proteins.
  • The vectors can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli.
  • Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity.
Please note that cloning into pASG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pASG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.
Specifications
Inducing Agent Inducible with anhydrotetracycline
Marker Gene Ampicillin
Promoter Tet
Product Type Vector
Content And Storage Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
Species E-coli
Protein Tag Twin-Strep-tag for N-terminal and FLAG-tag for C-terminal, FLAG-tag
Cloning Method Direct Cloning
Concentration 250 ng/μL
Quantity 5 μg
Vector pASG-IBA164
For Use With (Application) Vector for recombinant expression in E. coli