missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ E 64

Potent and irreversible cysteine protease inhibitor

246.00€ - 941.00€

Specifications

Purity >98%
Product Type E 64
Molecular Weight (g/mol) 357.41
Inhibitors E 64
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18745999
View Documents
Tocris Bioscience™
5208/10
10 mg
246.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
18790288
View Documents
Tocris Bioscience™
5208/50
50 mg
941.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potent and irreversible cysteine protease inhibitor (IC50 values are 1.4, 2.5 and 4.1 nM for cathepsin K, L and S respectively. Also inhibits papain, calpain, cathepsin B and cathepsin H. Has no effect on serine proteases. Reduces HSC-3 tongue cancer cell invasion and induces oxidative stress and apoptosis in filarial parasites. Also inhibits autophagy-associated lysosomal degradation in vitro.
Specifications

Specifications

>98%
E 64
357.41
E 64
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.