Learn More
JULABO™ DYNEO™ DD-201F Refrigerated/Heating Circulator
Circulators have a wide working temperature range. Suitable for both internal and external applications. Pump capacity is 22 L/min with pressure of 0.6 bar. The cooling machines operate precisely and reliably even at elevated ambient temperatures up to +40 °C.
Brand:  JULABO™ 9021702.D
 View more versions of this product
Product Code. 15910277
  • 5405.00€ / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Description
  • Powerful cooling machines
  • Suitable for internal and external applications
  • Optimized cooling coil design saves space in the bath tank
  • Powerful and infinitely adjustable pressure pump
  • Flow rate 22 L/min, pressure 0.6 bar
  • Easy switching between internal and external circulation
  • Large color TFT display, multilingual interface
  • Central rotary knob (controller) simplifies operation
  • Integrated programmer for 8 x 60 program steps
  • Integrated external Pt100 connection
  • USB port
  • RS232 interface or analog interfaces (optional)
  • Bath cover included with delivery
  • Integrated drain makes emptying liquid easy and safe
Specifications
Specifications
Refrigerated/Heating Circulator
0.2 kW
3.5 in. TFT Display
RS232 interface
0.1 to 0.6 bar
44 x 41 x 44 cm
24.7 kg
3 to 4 L
8 to 23 L/min.
2 kW
DYNEO™ DD-201F
-20°C to 200°C
15 cm
100 V/50-60 Hz, 115 V/60 Hz, 230 V/50-60 Hz