Få mere at vide
Få mere at vide
DWK Life Sciences™ DURAN™ Vigreux Column, with 2 standard ground joints, complete, with slide-on glass jacket
Shop alle DWK Life Sciences produkter
Klik for at se tilgængelige muligheder
Standard Taper Joint:
NS 24/29
NS 29/32
Length (Metric):
450 mm
650 mm
Pakningsstørrelse:
Each
Beskrivelse
Vigreux columns are used in fractionated distillation for improved separation of substances. The slightly downward angled indentations considerably increase the contact surface between the gas phase and the glass, i.e. the cooling surface.
- Tight closure due to standard ground joint
- With slide-on glass jacket
Tekniske data
Tekniske data
| For Use With (Application) | Fractional distillation; Laboratory coolers, condensation of gases and vapors |
| Length (Metric) | 650 mm |
| Material | Boro 3.3 |
| Quantity | 1 Ea. |
| Standard Taper Joint | NS 29/32 |
| Autoclavable | Autoclavable |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion