Learn More
DWK Life Sciences DURAN™ TILT Screw Cap, GL 56, White PP
DURAN™ TILT GL 56 screw cap features a secure grip, fast opening, and closing
Brand:  DWK Life Sciences 292295602
24.75 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 15583537
  • 28.00€ / Pack of 10
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Description
The ergonomic cap is easy to open and close. Made from non-cytotoxic materials. The wide mouth ensures safe and easy pipetting.
  • The ergonomic cap, made from non-cytotoxic materials, is easy to open and close
  • The wide GL 56 moth ensures safe and easy pipetting and helps avoid contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
Polypropylene
10/Pk.
Autoclavable
62 mm
Working with cell culture media under sterile conditions in biosafety cabinets and clean hoods; Pipetting liquids in biosafety cabinets and clean hoods
GL 56