Learn More
DWK Life Sciences DURAN™ Stopcock Key, hollow, threaded glass handle, ground, with three part retaining device
To be used for higher tightness requirements.
Brand:  DWK Life Sciences 286043007
 View more versions of this product
Product Code. 15864528
  • 1185.00€ / Pack of 50
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Hollow DURAN™ tap plugs are well suited for work with temperature changes and higher tightness requirements.
  • Hollow tap plug with thread, glass handle and 3-piece tap safety device
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2.5 mm
Autoclavable
50/Pk.
Borosilicate Glass (3.3), Polypropylene
14.5 NS
When working with temperature changes and higher tightness requirements