Learn More
DWK Life Sciences DURAN™ Stopcock Key, ground, with groove
For use with ground DURAN™ single way stopcocks.
Brand:  DWK Life Sciences 286036303
565.25 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15854518
  • 641.00€ / Pack of 25
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Description
DURAN™ tap plugs with groove are solid tap plugs which are secured by an O-ring.
  • With groove, polished
  • The groove serves to hold the O-ring, which secures the plug from falling out of the tap.
  • Solid glass tap plugs
  • Taper 1:10
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10.0 mm
Autoclavable
25/Pk.
Borosilicate Glass (3.3)
29.2 NS