Learn More
DWK Life Sciences DURAN™ Stopcock Key, ground, with groove
For use with ground DURAN™ single way stopcocks.
Brand: DWK Life Sciences 286038407
This item is not returnable.
View return policy
Description
DURAN™ tap plugs with groove are solid tap plugs which are secured by an O-ring.
- With groove, polished
- The groove serves to hold the O-ring, which secures the plug from falling out of the tap.
- Solid glass tap plugs
- Taper 1:10
Specifications
1.5 mm | |
Autoclavable | |
50/Pk. |
Borosilicate Glass (3.3) | |
10.0 NS |