Learn More
DWK Life Sciences DURAN™ Standard Cone with tip, finished
Core cut with extension.
3.08€ - 518.00€
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15911541
|
DWK Life Sciences
286166801 |
26 mm |
257.00€
Pack of 50 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15824588
|
DWK Life Sciences
286164406 |
13 mm |
166.00€
Pack of 50 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15844588
|
DWK Life Sciences
286165205 |
17 mm |
201.00€
Pack of 50 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15814588
|
DWK Life Sciences
|
11 mm |
205.00€
Pack of 50 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15874588
|
DWK Life Sciences
286168605 |
30 mm |
91.65€
Pack of 10 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15884588
|
DWK Life Sciences
|
40 mm |
126.00€
Pack of 10 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15804598
|
DWK Life Sciences
286170808 |
46 mm |
203.00€
Pack of 10 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15894578
|
DWK Life Sciences
286163007 |
6 mm |
74.65€
Pack of 10 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
10182121
|
DWK Life Sciences
286166004 |
22 mm | N/A | ||||||
10429732
|
DWK Life Sciences
286169601 |
90 mm |
111.00€
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15804588
|
DWK Life Sciences
286163401 |
8 mm |
241.00€
Pack of 50 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15894588
|
DWK Life Sciences
286170208 |
36 mm |
126.00€
Pack of 10 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15814598
|
DWK Life Sciences
286171007 |
50 mm |
234.00€
Pack of 10 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15824598
|
DWK Life Sciences
286171204 |
56 mm |
276.00€
Pack of 10 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15834598
|
DWK Life Sciences
286171607 |
65 mm |
518.00€
Pack of 10 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
10489152
|
DWK Life Sciences
286172209 |
80 mm |
79.55€
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
Description
The DURAN™ cores with outlet comply with DIN 12249 Form G. The process has the function of a drip ring. It prevents the liquid from running down the inner wall of the vessel. For this purpose, the outlet is usually cut at an angle and fused. At the same time, mediums do not come into contact with the greased ground surface. The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe.
- Ground (finer grinding quality, less roughness depth)
- The additional drain on the grinding enables a clean drain/leakage of liquids without wetting the grinding.
- Tight closure due to standard ground joint
- Glass articles for further processing