missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Single way Stopcock, ground and polished, without key

Ground and polished shut-off taps without plug for grease free operation.

Brand:  DWK Life Sciences 286439001

 View more versions of this product

Product Code. 15854758

  • 790.00€ / Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ground and polished DURAN™ single way stopcocks are shut-off taps which are combined with PTFE valve plugs to allow grease free operation.

  • Ground shut-off taps without tap plug
  • Fat-free working through the polished tap surface
  • Taper 1:10
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 mm
Borosilicate Glass (3.3)
12.5 NS
50/Pk.
8 mm
Autoclavable
1.5 mm
Coarse dosing of liquids; Installation in glass apparatus and devices
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions