Learn More
DWK Life Sciences DURAN™ Single way Stopcock, ground and polished, without key
Ground and polished shut-off taps without plug for grease free operation.
Brand:  DWK Life Sciences 286441007
 View more versions of this product
Product Code. 15824768
  • 173.00€ / Pack of 5
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Description
Ground and polished DURAN™ single way stopcocks are shut-off taps which are combined with PTFE valve plugs to allow grease free operation.
  • Ground shut-off taps without tap plug
  • Fat-free working through the polished tap surface
  • Taper 1:10
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6 mm
Borosilicate Glass (3.3)
21.5 NS
5/Pk.
13 mm
Autoclavable
1.5 mm
Coarse dosing of liquids; Installation in glass apparatus and devices