missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Single way Stopcock, complete with DURAN™ key with groove

Ground shut-off taps without tap plug.On-and-off taps with DURAN™ grooved tap plugs.

Brand:  DWK Life Sciences 286402402

168.10 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15864698

  • 177.00€ / Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

DURAN™ single way stopcocks complete with DURAN™ grooved tap plugs are shut-off taps in which the DURAN™ grooved tap plug is secured via the O-ring.

  • Ground shut-off valve sleeve complete with DURAN™ grooved plug
  • Securing of the tap plug by means of O-ring
  • Taper 1:10
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.5 mm
Borosilicate Glass (3.3)
10 NS
10/Pk.
7 mm
Autoclavable
1.5 mm
Installation in glass apparatus and devices
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions