Learn More
DWK Life Sciences DURAN™ Single way Stopcock, complete with DURAN™ key with groove
Ground shut-off taps without tap plug.On-and-off taps with DURAN™ grooved tap plugs.
Brand:  DWK Life Sciences 286407803
284.05 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15844708
  • 321.00€ / Pack of 5
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Description
DURAN™ single way stopcocks complete with DURAN™ grooved tap plugs are shut-off taps in which the DURAN™ grooved tap plug is secured via the O-ring.
  • Ground shut-off valve sleeve complete with DURAN™ grooved plug
  • Securing of the tap plug by means of O-ring
  • Taper 1:10
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mm
Borosilicate Glass (3.3)
29.2 NS
5/Pk.
18 mm
Autoclavable
2.5 mm
Installation in glass apparatus and devices