Learn More
DWK Life Sciences DURAN™ Multi-purpose Cylinder, with round base, without graduation
Cylinders without graduation can be used to mix components.
Brand: DWK Life Sciences 213985201
175.75 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility.
- Rough ground rim
- With round base
Specifications
Borosilicate Glass (3.3) | |
Diluting solutions; Mixing several components | |
Autoclavable |
40 mm | |
280 mL | |
10/Pk. |