Learn More
DWK Life Sciences DURAN™ Measuring Cylinder with UKAS Certificate, Class A
The DURAN™ Class A measuring cylinders with UKAS certificate are calibrated under defined conditions in accordance with ISO 1042, DIN 12664 and ISO 17025 standards.
167.00€ - 289.00€
Specifications
Material Borosilicate Glass (3.3)
For Use With (Application) Measuring liquid amounts; Diluting solutions; Mixing several components; Volumetric measurement
Includes 1/Pk.
Autoclavable Autoclavable
View More Specs
Products 9
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15806871
View Documents
DWK Life Sciences
213973602
250 mL
184.00€
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
15816871
View Documents
DWK Life Sciences
213974401
500 mL
203.00€
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
15826871
View Documents
DWK Life Sciences
213975406
1000 mL
230.00€
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
15836871
View Documents
DWK Life Sciences
213976308
2000 mL
289.00€
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
15856861
View Documents
DWK Life Sciences
213970708
5 mL
168.00€
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
15866861
View Documents
DWK Life Sciences
213970802
10 mL
167.00€
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
15876861
View Documents
DWK Life Sciences
213971404
25 mL
171.00€
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
15886861
View Documents
DWK Life Sciences
213971704
50 mL
171.00€
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
15896861
View Documents
DWK Life Sciences
213972409
100 mL
171.00€
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
Description
Description
Our DURAN™ volumetric measuring instruments already meet the highest accuracy standard. The DURAN™ range has now been expanded to include the product range of volumetric flasks and measuring cylinders with UKAS individual certificates. This new product line is tested and calibrated according to the requirements of the United Kingdom Accreditation Services (UKAS) and complies with ISO 17025, a standard for worldwide comparability of test and calibration results.
  • The volume content tolerances conform to accuracy Class A
  • High precision due to better visibility of the printed ring mark
  • The blue printed image is highly chemically resistant
  • Easy-to-read scale with graduation marks to estimate the content
  • Thanks to the exact processing and precisely calibrated scales, they permit the highly accurate determination and measurements of volume
  • Maximum minimization of unwanted influences
Specifications
Specifications
Borosilicate Glass (3.3)
Measuring liquid amounts; Diluting solutions; Mixing several components; Volumetric measurement
1/Pk.