Learn More
Karl Hecht™ DURAN™ Ground Joint Erlenmeyer Flasks
Provided with a reinforced standard ground neck with joint size clearly and durably marked.
Brand: Karl Hecht™ 41915034
This item is not returnable.
View return policy
Description
- Graduated with durable white markings
- Blank label area included
- Manufactured from DURAN™ borosilicate glass 3.3
- Stoppers sold separately
Specifications
Clear | |
Borosilicate Glass (3.3) | |
45/40 | |
Yes |
250 mL | |
Reinforced | |
White | |
Flat |