Learn More
Hirschmann™ DURAN™ Test Tubes with Blue Graduations
Click to view available options
Capacity (Metric):
10 mL
20 mL
25 mL
Graduations:
0.1 mL
0.2 mL
0.5 mL
Unit Size:
Pack of 10
Description
- With spout for easy and spill-free pouring
- Round base
- Clear glass for excellent clarity
- Available in a range of capacities
Specifications
Specifications
| Color | Clear |
| Capacity (Metric) | 25 mL |
| Length (Metric) | 215 mm |
| Material | Borosilicate Glass (3.3) |
| Includes | Spout |
| Quantity | 10/Pk. |
| Graduated | Yes |
| Diameter (Metric) | 18 mm |
| Product Type | Test Tube |
| Graduations | 0.5 mL |