Learn More
DWK Life Sciences DURAN™ Blank for reaction vessel
For processing as a reaction vessel.
Brand:  DWK Life Sciences 210208607
193.80 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10419152
  • 220.00€ / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DURAN™ blanks for reaction vessels are convincing due to their good wall thickness distribution and enable the cost-effective production of reaction vessels with reproducible geometry. For pressure resistance, please refer to our notes in the technical appendix of the laboratory glassware catalogue. For glass articles which deviate from the original form due to further processing, the permissible operating data must be determined in accordance with the technical rules and regulations.
  • Component for further processing into reaction vessels
  • Due to the geometry and the massive wall thickness, the blanks are suitable for devices with pressure and vacuum applications.
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
240 mm
Borosilicate Glass (3.3)
1/Pk.
10,000 mL
For processing as a reaction vessel
Autoclavable