Learn More
WTW™ DURAN™ 500mL Measuring Bottle
For respirometric measurements of BSB, soil respiration, degradability tests and biogas examinations.
Brand: WTW™ 209111
This item is not returnable.
View return policy
Description
- Tested for respective applications
- Supports accurate measurements
- Manufactured from durable DURAN glass
Specifications
500mL | |
Measuring Bottle | |
Respirometric measurements, soil respiration, degradability tests, biogas examinations | |
DURAN™ Glass |