missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DTNB; Ellman's Reagent, 5,5'-Dithiobis-(2-Nitrobenzoic Acid)

Product Code. 11594726
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 11594726

Brand: Invitrogen™ D8451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DTNB; Ellman's Reagent, 5,5'-Dithiobis-(2-Nitrobenzoic Acid)

DTNB or Elman's reagent can be used to quantitate thiols in proteins, cells and plasma by absorption measurements. It readily forms a mixed disulfide with thiols, liberating the chromophore 5-merapto-2-nitrobenzoic acid (absorption maximum 410 nm). Only protein thiols that are accessible to this water-soluble reagent are modified.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Elman's Reagent
Quantity 10 g

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.